Product Details

SNP ID
rs150002386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6328856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCTTGATGTTCAGGTCTCCAA[C/G]ACCTATGTGAAACAGAAGACATCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TPD52L3 PubMed Links

Gene Details

Gene
TPD52L3
Gene Name
tumor protein D52-like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001874.2 508 Missense Mutation AAC,AAG N87K NP_001001874.1
NM_001001875.3 508 Missense Mutation AAC,AAG N87K NP_001001875.1
NM_033516.5 508 Missense Mutation AAC,AAG N87K NP_277051.3
XM_017015280.1 508 Missense Mutation AAC,AAG N87K XP_016870769.1

View Full Product Details