Product Details

SNP ID
rs150457993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:70386359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTAGATACAAGATATATTAGTG[A/G]TGAGGCTATTGATGTAAAAGATACA
Phenotype
MIM: 602902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLF9 PubMed Links

Gene Details

Gene
KLF9
Gene Name
Kruppel like factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206.2 3446 UTR 3 NP_001197.1

View Full Product Details