Product Details

SNP ID
rs150479898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120817915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCTCTAGAAGAAATCATTCGGC[C/T]CCTTCTACTGCTGTCGTGGAAACAG
Phenotype
MIM: 604452
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMD5 PubMed Links

Gene Details

Gene
PSMD5
Gene Name
proteasome 26S subunit, non-ATPase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270427.1 1618 Silent Mutation GGA,GGG G459G NP_001257356.1
NM_005047.3 1618 Silent Mutation GGA,GGG G502G NP_005038.1
XM_011518866.2 1618 Intron XP_011517168.1
XM_017014927.1 1618 Intron XP_016870416.1
XM_017014928.1 1618 Silent Mutation GGA,GGG G288G XP_016870417.1

View Full Product Details