Product Details

SNP ID
rs150946607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34254466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGCTCCTCAGACTCTGGATAC[A/G]CTTGGATTTCAGAACCATGATTTCA
Phenotype
MIM: 613747 MIM: 609787
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF24 PubMed Links

Gene Details

Gene
KIF24
Gene Name
kinesin family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194313.2 2745 Missense Mutation CGT,TGT R1341C NP_919289.2
XM_005251451.2 2745 Missense Mutation CGT,TGT R980C XP_005251508.1
XM_006716764.2 2745 Missense Mutation CGT,TGT R883C XP_006716827.1
XM_011517861.1 2745 Missense Mutation CGT,TGT R1372C XP_011516163.1
XM_011517863.2 2745 Missense Mutation CGT,TGT R1341C XP_011516165.1
XM_017014697.1 2745 Missense Mutation CGT,TGT R1341C XP_016870186.1
XM_017014698.1 2745 Missense Mutation CGT,TGT R1052C XP_016870187.1
Gene
UBAP1
Gene Name
ubiquitin associated protein 1
There are no transcripts associated with this gene.

View Full Product Details