Product Details

SNP ID
rs150968065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135812354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTACAAAAAGGAATGGAGGTCC[A/G]GTACACACTACACTGAGAAACCTCA
Phenotype
MIM: 613774
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMSAP1 PubMed Links

Gene Details

Gene
CAMSAP1
Gene Name
calmodulin regulated spectrin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015447.3 Intron NP_056262.3
XM_005263396.3 Intron XP_005263453.1
XM_005263397.1 Intron XP_005263454.1
XM_011518272.1 Intron XP_011516574.1
XM_011518273.2 Intron XP_011516575.1
XM_011518274.1 Intron XP_011516576.1
XM_017014301.1 Intron XP_016869790.1
XM_017014302.1 Intron XP_016869791.1

View Full Product Details