Product Details

SNP ID
rs104894737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:11294790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACATTTCAGAACCATCAAGAAA[C/T]GGGGACCTGGATTTTATTTGCCTGC
Phenotype
MIM: 300391 MIM: 300118
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMELX PubMed Links

Gene Details

Gene
AMELX
Gene Name
amelogenin, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142.2 1171 Missense Mutation ACG,ATG T1M NP_001133.1
NM_182680.1 1171 Missense Mutation ACG,ATG T1M NP_872621.1
NM_182681.1 1171 Missense Mutation ACG,ATG T1M NP_872622.1
XM_017029404.1 1171 Missense Mutation ACG,ATG T1M XP_016884893.1
Gene
ARHGAP6
Gene Name
Rho GTPase activating protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287242.1 1171 Intron NP_001274171.1
NM_006125.2 1171 Intron NP_006116.2
NM_013423.2 1171 Intron NP_038267.1
NM_013427.2 1171 Intron NP_038286.2

View Full Product Details