Product Details

SNP ID
rs137929349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49242910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGACTCAGCTATTCTCCTCCGGG[C/T]CATTGGGAGCCAAATTCGGGACCAG
Phenotype
MIM: 300110 MIM: 300859 MIM: 300292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNA1F PubMed Links

Gene Details

Gene
CACNA1F
Gene Name
calcium voltage-gated channel subunit alpha1 F
There are no transcripts associated with this gene.

Gene
CCDC22
Gene Name
coiled-coil domain containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014008.4 556 Missense Mutation GCC,GTC A129V NP_054727.1
XM_005272599.3 556 Missense Mutation GCC,GTC A129V XP_005272656.1
Gene
FOXP3
Gene Name
forkhead box P3
There are no transcripts associated with this gene.

View Full Product Details