Product Details

SNP ID
rs138083924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130215326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCAATGCACTTGTGAATTCCCC[A/G]ACGACACCTTATGGAGACGGAAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF280C PubMed Links

Gene Details

Gene
ZNF280C
Gene Name
zinc finger protein 280C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017666.4 2049 Missense Mutation NP_060136.1
XM_006724765.3 2049 Missense Mutation XP_006724828.1

View Full Product Details