Product Details

SNP ID
rs138345294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78113989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTTTAACTTTCATACTGCTCA[A/G]GTGTCTTCATCTCTTCCTCTTCTCA
Phenotype
MIM: 311800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGK1 PubMed Links

Gene Details

Gene
PGK1
Gene Name
phosphoglycerate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000291.3 Intron NP_000282.1

View Full Product Details