Product Details

SNP ID
rs138947407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134789106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCAGGCTCAAGCTCTCATCCCA[C/T]GATTGGCTTATCTGCATTGCCGTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM122B PubMed Links

Gene Details

Gene
FAM122B
Gene Name
family with sequence similarity 122B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166599.2 1330 Silent Mutation TCA,TCG S85S NP_001160071.1
NM_001166600.2 1330 Silent Mutation TCA,TCG S104S NP_001160072.1
NM_001170756.1 1330 Silent Mutation TCA,TCG S104S NP_001164227.1
NM_001170757.1 1330 Silent Mutation TCA,TCG S51S NP_001164228.1
NM_145284.5 1330 Silent Mutation TCA,TCG S85S NP_660327.2
XM_006724730.2 1330 Silent Mutation TCA,TCG S104S XP_006724793.1
XM_006724732.2 1330 Silent Mutation TCA,TCG S104S XP_006724795.1
XM_006724733.3 1330 Silent Mutation TCA,TCG S85S XP_006724796.1
XM_011531282.1 1330 Silent Mutation TCA,TCG S108S XP_011529584.1
XM_011531283.1 1330 Silent Mutation TCA,TCG S108S XP_011529585.1
XM_011531284.1 1330 Silent Mutation TCA,TCG S104S XP_011529586.1
XM_011531285.1 1330 Silent Mutation TCA,TCG S104S XP_011529587.1
XM_011531286.1 1330 Silent Mutation TCA,TCG S89S XP_011529588.1
XM_011531287.1 1330 Silent Mutation TCA,TCG S108S XP_011529589.1
XM_011531288.1 1330 Silent Mutation TCA,TCG S85S XP_011529590.1
XM_011531289.1 1330 Silent Mutation TCA,TCG S108S XP_011529591.1
XM_011531290.1 1330 Silent Mutation TCA,TCG S85S XP_011529592.1
XM_011531291.1 1330 Silent Mutation TCA,TCG S108S XP_011529593.1
XM_011531292.1 1330 Silent Mutation TCA,TCG S108S XP_011529594.1
XM_011531293.1 1330 Silent Mutation TCA,TCG S104S XP_011529595.1
XM_011531294.1 1330 Silent Mutation TCA,TCG S85S XP_011529596.1
XM_017029306.1 1330 Silent Mutation TCA,TCG S89S XP_016884795.1
XM_017029307.1 1330 Silent Mutation TCA,TCG S89S XP_016884796.1
Gene
FAM122C
Gene Name
family with sequence similarity 122C
There are no transcripts associated with this gene.

View Full Product Details