Product Details

SNP ID
rs139106055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49057936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTCGTAAGGACTCAAAGCTCTCA[C/T]TTCCTTCTCTGGCATGAAGCCTGAC
Phenotype
MIM: 300947
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC120 PubMed Links

Gene Details

Gene
CCDC120
Gene Name
coiled-coil domain containing 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163321.2 Intron NP_001156793.2
NM_001163322.2 Intron NP_001156794.1
NM_001163323.2 Intron NP_001156795.1
NM_001271835.1 Intron NP_001258764.1
NM_001271836.1 Intron NP_001258765.1
NM_033626.3 Intron NP_296375.1

View Full Product Details