Product Details

SNP ID
rs139134641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:43950026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCGCACCTGGCCAGGAGCACCA[G/T]CTGGGGAAAGAAAAGGAGGTGGTTA
Phenotype
MIM: 300658
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NDP PubMed Links

Gene Details

Gene
NDP
Gene Name
NDP, norrin cystine knot growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000266.3 754 Missense Mutation ATG,CTG M59L NP_000257.1

View Full Product Details