Product Details
- SNP ID
-
rs139518179
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:10033526 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACGTGGCATATTGACAAAAATGTC[A/G]AGGATTACCAAGAATAGCTTCTTGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WWC3
PubMed Links
Gene Details
- Gene
- WWC3
- Gene Name
- WWC family member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015691.3 |
|
Intron |
|
|
NP_056506.2 |
View Full Product Details