Product Details
- SNP ID
-
rs140532942
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:74740352 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGATCACCTATTGCTGTTTTCCCA[C/T]TGCTTTTGCTCACACCCTTGATTGA
- Phenotype
-
MIM: 300524
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA2022
PubMed Links
Gene Details
- Gene
- KIAA2022
- Gene Name
- KIAA2022
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001008537.2 |
4822 |
Missense Mutation |
AAT,AGT |
N1402S |
NP_001008537.1 |
View Full Product Details