Product Details

SNP ID
rs140734874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49071995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGTTGCGCAGGCGCAACGAGGC[A/G]TGCACCAGGATCACTGTGGGCGGCA
Phenotype
MIM: 300947 MIM: 300840 MIM: 300526
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC120 PubMed Links

Gene Details

Gene
CCDC120
Gene Name
coiled-coil domain containing 120
There are no transcripts associated with this gene.

Gene
PRAF2
Gene Name
PRA1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007213.2 469 Silent Mutation CAC,CAT H137H NP_009144.1
Gene
WDR45
Gene Name
WD repeat domain 45
There are no transcripts associated with this gene.

View Full Product Details