Product Details

SNP ID
rs141880369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50592243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTCAGTACTTCCCATGCTGTA[A/G]TATGTTCATGAATCTCCTGGAAATC
Phenotype
MIM: 300579
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHROOM4 PubMed Links

Gene Details

Gene
SHROOM4
Gene Name
shroom family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020717.3 8960 UTR 3 NP_065768.2
XM_017029682.1 8960 Intron XP_016885171.1
XM_017029683.1 8960 Intron XP_016885172.1
XM_017029684.1 8960 Intron XP_016885173.1
XM_017029685.1 8960 Intron XP_016885174.1
XM_017029686.1 8960 Intron XP_016885175.1
XM_017029687.1 8960 Intron XP_016885176.1

View Full Product Details