Product Details

SNP ID
rs142033679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15287955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAGCGCACTTTTGCCCTGAGCA[G/T]TTCTACGCTTCAGGTTAGCTCCATA
Phenotype
MIM: 300626
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASB11 PubMed Links

Gene Details

Gene
ASB11
Gene Name
ankyrin repeat and SOCS box containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012428.2 741 Missense Mutation AAT,ACT N237T NP_001012428.1
NM_001201583.1 741 Missense Mutation AAT,ACT N241T NP_001188512.1
NM_080873.2 741 Missense Mutation AAT,ACT N258T NP_543149.1

View Full Product Details