Product Details

SNP ID
rs142471926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:141907005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCATTCTCCTCTCCAGATTGTTC[C/G]AAGTCTTCCTGAGTGGGAGGACTCC
Phenotype
MIM: 300223 MIM: 300469
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAGEC1 PubMed Links

Gene Details

Gene
MAGEC1
Gene Name
MAGE family member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005462.4 1925 Missense Mutation CCA,CGA P534R NP_005453.2
XM_011531418.2 1925 Missense Mutation CCA,CGA P534R XP_011529720.1
Gene
MAGEC3
Gene Name
MAGE family member C3
There are no transcripts associated with this gene.

View Full Product Details