Product Details

SNP ID
rs142507863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70269391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCAACTATATAGCCAACATGTC[A/C]AAGTAAGAATTCCTTCTAATCCTCT
Phenotype
MIM: 301770 MIM: 300038
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARR3 PubMed Links

Gene Details

Gene
ARR3
Gene Name
arrestin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004312.2 74 Silent Mutation TCA,TCC S2S NP_004303.2
XM_017029518.1 74 Intron XP_016885007.1
Gene
P2RY4
Gene Name
pyrimidinergic receptor P2Y4
There are no transcripts associated with this gene.

View Full Product Details