Product Details

SNP ID
rs143665198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:129788511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCTGGGGAAGAAGTGGAGGGC[A/G]GTGATTTCCCGAACCATGAACAGGA
Phenotype
MIM: 300441
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SASH3 PubMed Links

Gene Details

Gene
SASH3
Gene Name
SAM and SH3 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018990.3 416 Silent Mutation GCA,GCG A78A NP_061863.1
XM_006724763.1 416 Silent Mutation GCA,GCG A78A XP_006724826.1

View Full Product Details