Product Details

SNP ID
rs143934702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:116462833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCAAGGCACACAGAATGCTGCT[C/T]GCTAGGAGTAAATAGAAGTTCATGT
Phenotype
MIM: 300625 MIM: 300444
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CT83 PubMed Links

Gene Details

Gene
CT83
Gene Name
cancer/testis antigen 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017978.3 Intron NP_001017978.1
Gene
SLC6A14
Gene Name
solute carrier family 6 member 14
There are no transcripts associated with this gene.

View Full Product Details