Product Details

SNP ID
rs144035929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152115293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGGTTGCTGGAGCCCTTAATG[G/T]ATTGCCTCCATAGAGTGAAATCGAT
Phenotype
MIM: 300340
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAGEA10-MAGEA5 PubMed Links

Gene Details

Gene
MAGEA10-MAGEA5
Gene Name
MAGEA10-MAGEA5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204811.2 Intron NP_001191740.1
Gene
MAGEA5
Gene Name
MAGE family member A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021049.4 Intron NP_066387.1

View Full Product Details