Product Details

SNP ID
rs144060196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101397838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAGTCTTTTAATGACATCTGCA[C/T]TGTATTTTCTAGCTGAAGCAAAACA
Phenotype
MIM: 300300 MIM: 300644 MIM: 300902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTK PubMed Links

Gene Details

Gene
BTK
Gene Name
Bruton tyrosine kinase
There are no transcripts associated with this gene.

Gene
GLA
Gene Name
galactosidase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000169.2 1371 Missense Mutation ATG,GTG M421V NP_000160.1
Gene
RPL36A
Gene Name
ribosomal protein L36a
There are no transcripts associated with this gene.

Gene
RPL36A-HNRNPH2
Gene Name
RPL36A-HNRNPH2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199973.1 1371 Intron NP_001186902.1
NM_001199974.1 1371 Intron NP_001186903.1

View Full Product Details