Product Details

SNP ID
rs144206056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49176907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCTAGAGGGTTAGCTCACCTGG[C/T]GCTAACTCTGTGCTGGTGCCTTTGG
Phenotype
MIM: 300112 MIM: 300111
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAGIX PubMed Links

Gene Details

Gene
MAGIX
Gene Name
MAGI family member, X-linked
There are no transcripts associated with this gene.

Gene
PLP2
Gene Name
proteolipid protein 2
There are no transcripts associated with this gene.

Gene
PRICKLE3
Gene Name
prickle planar cell polarity protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307979.1 1354 Silent Mutation GCA,GCG A349A NP_001294908.1
NM_006150.4 1354 Silent Mutation GCA,GCG A417A NP_006141.2

View Full Product Details