Product Details

SNP ID
rs1003961315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48975236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCATCTCCCGAAGGTTCTCGTC[A/G]CCTGGCTTCACTAGGTCTCTCAGGA
Phenotype
MIM: 300408 MIM: 300281
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIPAP1 PubMed Links

Gene Details

Gene
GRIPAP1
Gene Name
GRIP1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020137.4 2387 Silent Mutation GGC,GGT G784G NP_064522.3
Gene
KCND1
Gene Name
potassium voltage-gated channel subfamily D member 1
There are no transcripts associated with this gene.

View Full Product Details