Product Details
- SNP ID
-
rs145290002
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:153670810 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACAGAGCCTAAGATGTCCCTGTC[A/G]AAGGCTGTGTCCCCAGAGATCCTGG
- Phenotype
-
MIM: 300680
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PNCK
PubMed Links
Gene Details
- Gene
- PNCK
- Gene Name
- pregnancy up-regulated nonubiquitous CaM kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039582.3 |
876 |
Silent Mutation |
TTC,TTT |
F359F |
NP_001034671.3 |
NM_001135740.1 |
876 |
Silent Mutation |
TTC,TTT |
F293F |
NP_001129212.1 |
XM_005274651.2 |
876 |
Silent Mutation |
TTC,TTT |
F276F |
XP_005274708.1 |
XM_006724809.3 |
876 |
Silent Mutation |
TTC,TTT |
F276F |
XP_006724872.1 |
XM_011531107.2 |
876 |
Silent Mutation |
TTC,TTT |
F276F |
XP_011529409.2 |
XM_011531108.2 |
876 |
Silent Mutation |
TTC,TTT |
F294F |
XP_011529410.1 |
XM_011531110.2 |
876 |
Silent Mutation |
TTC,TTT |
F276F |
XP_011529412.1 |
XM_011531111.2 |
876 |
Silent Mutation |
TTC,TTT |
F276F |
XP_011529413.1 |
XM_011531112.2 |
876 |
Silent Mutation |
TTC,TTT |
F276F |
XP_011529414.1 |
XM_017029277.1 |
876 |
Silent Mutation |
TTC,TTT |
F359F |
XP_016884766.1 |
XM_017029278.1 |
876 |
Silent Mutation |
TTC,TTT |
F276F |
XP_016884767.1 |
XM_017029279.1 |
876 |
Silent Mutation |
TTC,TTT |
F276F |
XP_016884768.1 |
XM_017029280.1 |
876 |
Intron |
|
|
XP_016884769.1 |
View Full Product Details