Product Details

SNP ID
rs145654350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72186524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAAGGTCCCAAAGGTGGTGGCA[A/G]TGCAGTAAAGGTGAGTTACTGGTTC
Phenotype
MIM: 300252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIN4 PubMed Links

Gene Details

Gene
PIN4
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170747.1 217 Missense Mutation AAT,AGT N61S NP_001164218.1
NM_006223.3 217 Missense Mutation AAT,AGT N61S NP_006214.2

View Full Product Details