Product Details
- SNP ID
-
rs145771147
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:16810537 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAGTCCTGTTGGAACTGGGCTTA[C/T]TAATAAACAAAAAGATTTTGTAGAG
- Phenotype
-
MIM: 300677
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TXLNG
PubMed Links
Gene Details
- Gene
- TXLNG
- Gene Name
- taxilin gamma
View Full Product Details