Product Details

SNP ID
rs146126448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:129745959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGCCCCATCTTTCCTGAGAGC[A/G]CCTACATGCAGCCAAAAGTTGACCT
Phenotype
MIM: 300145
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XPNPEP2 PubMed Links

Gene Details

Gene
XPNPEP2
Gene Name
X-prolyl aminopeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003399.5 Intron NP_003390.4

View Full Product Details