Product Details

SNP ID
rs146203325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153592529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCCACCACGGCTTCTCAGCCT[C/T]GACCTCGGCGGGCACCTCAACTCCG
Phenotype
MIM: 300014 MIM: 300708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP2B3 PubMed Links

Gene Details

Gene
ATP2B3
Gene Name
ATPase plasma membrane Ca2+ transporting 3
There are no transcripts associated with this gene.

Gene
FAM58A
Gene Name
family with sequence similarity 58 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130997.2 1262 Intron NP_001124469.1
NM_152274.4 1262 Intron NP_689487.2
XM_005277920.4 1262 Intron XP_005277977.1
XM_005277921.4 1262 Intron XP_005277978.1
XM_011531214.2 1262 Missense Mutation AAG,GAG K170E XP_011529516.1
XM_011531215.2 1262 Missense Mutation AAG,GAG K170E XP_011529517.1
Gene
LOC105373383
Gene Name
uncharacterized LOC105373383
There are no transcripts associated with this gene.

View Full Product Details