Product Details

SNP ID
rs147158965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:37010791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCACCTGGAGCCTCCCAAGACTC[A/G]TCGAGTGTCCAGTCTCCGCCTGGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM47C PubMed Links

Gene Details

Gene
FAM47C
Gene Name
family with sequence similarity 47 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013736.2 Intron NP_001013758.1

View Full Product Details