Product Details

SNP ID
rs147232392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137566740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCAGTTCCCTGGGCTGGGAGTG[G/T]GCAGCTTCGGCGCGCCGCGCCACCA
Phenotype
MIM: 300265
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZIC3 PubMed Links

Gene Details

Gene
ZIC3
Gene Name
Zic family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003413.3 554 Missense Mutation GGC,TGC G17C NP_003404.1
XM_017029802.1 554 Missense Mutation GGC,TGC G17C XP_016885291.1

View Full Product Details