Product Details
- SNP ID
-
rs148855291
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:101841748 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCAGCACTAGCAACCTGGACAA[A/G]GAAGCATGCCCGATTTCGGATGTAG
- Phenotype
-
MIM: 300319
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NXF5
PubMed Links
Gene Details
- Gene
- NXF5
- Gene Name
- nuclear RNA export factor 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032946.2 |
526 |
Missense Mutation |
CTT,TTT |
L56F |
NP_116564.2 |
View Full Product Details