Product Details

SNP ID
hCV175596426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154446528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGAGGCGAGGAGGAAGAGGAGG[C/T]GGCCATGTATGAGGAGGAGATGGAA
Phenotype
MIM: 300453 MIM: 300104
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM50A PubMed Links

Gene Details

Gene
FAM50A
Gene Name
family with sequence similarity 50 member A
There are no transcripts associated with this gene.

Gene
GDI1
Gene Name
GDP dissociation inhibitor 1
There are no transcripts associated with this gene.

Gene
MIR6858
Gene Name
microRNA 6858
There are no transcripts associated with this gene.

View Full Product Details