Product Details

SNP ID
rs149821108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:69505958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGTAGCCTGGACACCCTGATG[G/T]GGGACCTGCTGGCCGTGGTGGCCAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM155B PubMed Links

Gene Details

Gene
FAM155B
Gene Name
family with sequence similarity 155 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015686.2 724 Missense Mutation GGG,TGG G226W NP_056501.2
XM_011530908.2 724 Missense Mutation GGG,TGG G226W XP_011529210.1
XM_011530909.2 724 Missense Mutation GGG,TGG G226W XP_011529211.1

View Full Product Details