Product Details

SNP ID
rs149890335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:52862633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCTTCATTTAAACCTGTGGTTG[C/T]CTGTCACCTATAATAGAAATATAAC
Phenotype
MIM: 300740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XAGE3 PubMed Links

Gene Details

Gene
XAGE3
Gene Name
X antigen family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130776.1 441 Intron NP_570132.1
NM_133179.2 441 Silent Mutation AGA,AGG R107R NP_573440.1
XM_011530768.2 441 Silent Mutation AGA,AGG R107R XP_011529070.1
XM_011530769.2 441 Silent Mutation AGA,AGG R107R XP_011529071.1
XM_011530770.2 441 Silent Mutation AGA,AGG R107R XP_011529072.1

View Full Product Details