Product Details

SNP ID
rs150401178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:131060069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATTGACTTCCATGGTCCTCCTT[C/T]CTAGGAAACAGTGTCAGCTCTCCCC
Phenotype
MIM: 300937
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP36 PubMed Links

Gene Details

Gene
ARHGAP36
Gene Name
Rho GTPase activating protein 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282607.1 Intron NP_001269536.1
NM_144967.3 Intron NP_659404.2
XM_006724729.1 Intron XP_006724792.1
XM_011531280.1 Intron XP_011529582.1
Gene
LINC01201
Gene Name
long intergenic non-protein coding RNA 1201
There are no transcripts associated with this gene.

View Full Product Details