Product Details

SNP ID
rs150684784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:26559849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTCACTTAAAAATTATTTGACT[A/G]TTGACAGGTTATTAAATCTCTGTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VENTXP1 PubMed Links

Gene Details

Gene
VENTXP1
Gene Name
VENT homeobox pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details