Product Details

SNP ID
rs151108723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:65518320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACGCTCCAATACAGGCTGTCTA[A/G]TGTATATGGAGACTTCCCAATGGGG
Phenotype
MIM: 300964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAS1L PubMed Links

Gene Details

Gene
LAS1L
Gene Name
LAS1 like, ribosome biogenesis factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170649.1 1619 Silent Mutation CTA,TTA L515L NP_001164120.1
NM_001170650.1 1619 Silent Mutation CTA,TTA L473L NP_001164121.1
NM_031206.4 1619 Silent Mutation CTA,TTA L532L NP_112483.1
XM_005262301.1 1619 Silent Mutation CTA,TTA L532L XP_005262358.1
XM_005262304.1 1619 Intron XP_005262361.1
XM_005262305.3 1619 Intron XP_005262362.1
XM_005262306.3 1619 Intron XP_005262363.1
XM_011531045.1 1619 Silent Mutation CTA,TTA L490L XP_011529347.1
XM_011531046.1 1619 Intron XP_011529348.1
XM_017029877.1 1619 Silent Mutation CTA,TTA L515L XP_016885366.1
XM_017029878.1 1619 Intron XP_016885367.1
XM_017029879.1 1619 Silent Mutation CTA,TTA L230L XP_016885368.1
XM_017029880.1 1619 Silent Mutation CTA,TTA L213L XP_016885369.1

View Full Product Details