Product Details

SNP ID
rs1059528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:77721383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGTGTCCTTGACGGCAGCAAA[G/C]ACAAAGCGGATATTCTCGGTGTCTG
Phenotype
MIM: 600998
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
GNAQ PubMed Links

Gene Details

Gene
GNAQ
Gene Name
G protein subunit alpha q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002072.4 1234 Silent Mutation GTC,GTG V340V NP_002063.2
XM_017014628.1 1234 Silent Mutation GTC,GTG V282V XP_016870117.1

View Full Product Details