Product Details
- SNP ID
-
hCV175675638
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:230897 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGAGGCTACGGGCGCACCTACTT[C/G]AGCTGCACGTCTGCCCACACCAGCA
- Phenotype
-
MIM: 600857
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SDHA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2288459,rs2288461] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SDHA
- Gene Name
- succinate dehydrogenase complex flavoprotein subunit A
There are no transcripts associated with this gene.
View Full Product Details