Product Details

SNP ID
rs11547273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59697939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACAGCCAAGCCGAGTGTACTGG[T/G]ATGAGCCAAATATTCCATAACCAAG
Phenotype
MIM: 118955 MIM: 608625 MIM: 611753
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
CLTC PubMed Links

Gene Details

Gene
CLTC
Gene Name
clathrin heavy chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288653.1 122 Intron NP_001275582.1
NM_004859.3 122 Intron NP_004850.1
XM_005257012.3 122 Intron XP_005257069.1
Gene
PTRH2
Gene Name
peptidyl-tRNA hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015509.2 122 Missense Mutation ACC,CCC T15P NP_001015509.1
NM_016077.4 122 Missense Mutation ACC,CCC T14P NP_057161.1
XM_011524887.1 122 Missense Mutation ACC,CCC T14P XP_011523189.1
Gene
VMP1
Gene Name
vacuole membrane protein 1
There are no transcripts associated with this gene.

View Full Product Details