Product Details

SNP ID
rs11552431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1773023 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGTCTCGTAGTCCACCGCGTAGA[T/G]CTGGGAGTCGCGCGGCCTGTTCAGT
Phenotype
MIM: 610886 MIM: 605431 MIM: 611994 MIM: 601817 MIM: 610779 MIM: 611659
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
EME2 PubMed Links

Gene Details

Gene
EME2
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257370.1 117 Intron NP_001244299.1
XM_017023025.1 117 Intron XP_016878514.1
XM_017023026.1 117 Intron XP_016878515.1
Gene
MAPK8IP3
Gene Name
mitogen-activated protein kinase 8 interacting protein 3
There are no transcripts associated with this gene.

Gene
MRPS34
Gene Name
mitochondrial ribosomal protein S34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300900.1 117 Missense Mutation ATC,CTC I33L NP_001287829.1
NM_023936.1 117 Missense Mutation ATC,CTC I33L NP_076425.1
XM_017023595.1 117 Missense Mutation ATC,CTC I33L XP_016879084.1
Gene
NME3
Gene Name
NME/NM23 nucleoside diphosphate kinase 3
There are no transcripts associated with this gene.

Gene
NUBP2
Gene Name
nucleotide binding protein 2
There are no transcripts associated with this gene.

Gene
SPSB3
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
There are no transcripts associated with this gene.

View Full Product Details