Product Details

SNP ID
rs11553732
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:2185284 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCCTGCCGAGGGGGCTGTCATGC[A/G]GTTTCCAAGGTGCACATTTTCCACG
Phenotype
MIM: 615183 MIM: 176982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAAP20 PubMed Links

Gene Details

Gene
FAAP20
Gene Name
Fanconi anemia core complex associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146310.1 2123 Silent Mutation ACC,ACT T229T NP_001139782.1
NM_001256945.1 2123 Silent Mutation ACC,ACT T188T NP_001243874.1
NM_001256946.1 2123 Intron NP_001243875.1
NM_001256947.1 2123 Intron NP_001243876.1
NM_001282670.1 2123 Intron NP_001269599.1
NM_001282671.1 2123 Silent Mutation ACC,ACT T146T NP_001269600.1
NM_001282672.1 2123 Silent Mutation ACC,ACT T146T NP_001269601.1
NM_001282673.1 2123 UTR 3 NP_001269602.1
NM_182533.2 2123 Intron NP_872339.2
XM_006710419.3 2123 Intron XP_006710482.1
XM_006710421.3 2123 Intron XP_006710484.1
XM_011540914.2 2123 Intron XP_011539216.1
XM_011540921.2 2123 Intron XP_011539223.1
XM_011540922.1 2123 Intron XP_011539224.1
XM_017000553.1 2123 Intron XP_016856042.1
XM_017000554.1 2123 UTR 3 XP_016856043.1
XM_017000555.1 2123 Silent Mutation ACC,ACT T146T XP_016856044.1
XM_017000556.1 2123 Silent Mutation ACC,ACT T146T XP_016856045.1
XM_017000557.1 2123 Intron XP_016856046.1
Gene
LOC100506504
Gene Name
uncharacterized LOC100506504
There are no transcripts associated with this gene.

Gene
PRKCZ
Gene Name
protein kinase C zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033581.1 2123 UTR 3 NP_001028753.1
NM_001033582.1 2123 UTR 3 NP_001028754.1
NM_001242874.1 2123 UTR 3 NP_001229803.1
NM_002744.4 2123 UTR 3 NP_002735.3
XM_011541773.1 2123 Intron XP_011540075.1
XM_011541774.1 2123 Intron XP_011540076.1
XM_011541775.2 2123 Intron XP_011540077.1
XM_011541776.1 2123 Intron XP_011540078.1
XM_011541778.1 2123 Intron XP_011540080.1
XM_017001789.1 2123 Intron XP_016857278.1
XM_017001790.1 2123 UTR 3 XP_016857279.1
XM_017001791.1 2123 UTR 3 XP_016857280.1
XM_017001792.1 2123 Intron XP_016857281.1
XM_017001793.1 2123 Intron XP_016857282.1
XM_017001794.1 2123 Intron XP_016857283.1
XM_017001795.1 2123 Intron XP_016857284.1
XM_017001796.1 2123 UTR 3 XP_016857285.1
XM_017001797.1 2123 Intron XP_016857286.1
XM_017001798.1 2123 UTR 3 XP_016857287.1
XM_017001799.1 2123 UTR 3 XP_016857288.1
XM_017001800.1 2123 Intron XP_016857289.1
XM_017001801.1 2123 UTR 3 XP_016857290.1
XM_017001802.1 2123 Intron XP_016857291.1
XM_017001803.1 2123 Intron XP_016857292.1
XM_017001804.1 2123 UTR 3 XP_016857293.1
XM_017001805.1 2123 UTR 3 XP_016857294.1

View Full Product Details