Product Details

SNP ID
rs11557070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30445408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCTGGAAAGGTGGGGCTGGGG[T/C]CCGCTCCTGCCGGCAGGCCGGCTTC
Phenotype
MIM: 606218
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SEPHS2 PubMed Links

Gene Details

Gene
SEPHS2
Gene Name
selenophosphate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012248.3 568 Missense Mutation NP_036380.2

View Full Product Details