Product Details

SNP ID
rs11705099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:16964836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCTCTGCGTGGAGATGGGGTGT[G/T]GGGAGGAGCTGGAGTCAAAGGTGTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GAB4 PubMed Links

Gene Details

Gene
GAB4
Gene Name
GRB2 associated binding protein family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037814.1 804 Missense Mutation NP_001032903.1
XM_011546114.2 804 Missense Mutation XP_011544416.1
XM_011546115.1 804 Missense Mutation XP_011544417.1
XM_011546116.2 804 Intron XP_011544418.1
XM_011546117.1 804 Missense Mutation XP_011544419.1
XM_017028575.1 804 Missense Mutation XP_016884064.1
XM_017028576.1 804 Missense Mutation XP_016884065.1

View Full Product Details