Product Details
- SNP ID
-
rs11072
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
56
- Location
-
Chr.1:182574196 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTCATTATTCAATGAATATTTATT[G/A]AACATTCCCACTTGGCAGACTTTTT
- Phenotype
-
MIM: 180435
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNASEL
PubMed Links
Gene Details
- Gene
- RNASEL
- Gene Name
- ribonuclease L
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021133.3 |
3679 |
UTR 3 |
|
|
NP_066956.1 |
View Full Product Details