Product Details

SNP ID
rs11166385
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:99977906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCTAGAACCTTGTAATGTTAGT[A/G]CGTGGGGAGCAATTCTATCTTAATG
Phenotype
MIM: 605632
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35A3 PubMed Links

Gene Details

Gene
SLC35A3
Gene Name
solute carrier family 35 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271684.1 Intron NP_001258613.1
NM_001271685.1 Intron NP_001258614.1
NM_012243.2 Intron NP_036375.1
XM_005270691.4 Intron XP_005270748.1
XM_011541135.2 Intron XP_011539437.1
XM_011541136.2 Intron XP_011539438.1
XM_011541137.2 Intron XP_011539439.1
XM_011541138.2 Intron XP_011539440.1
XM_017000869.1 Intron XP_016856358.1
XM_017000870.1 Intron XP_016856359.1
XM_017000871.1 Intron XP_016856360.1
XM_017000872.1 Intron XP_016856361.1

View Full Product Details