Product Details

SNP ID
rs11538097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:2905361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGATGGACTGAGGGGAATAGTG[C/T]AAGGTCCTCCCATCCCCAGTATTGG
Phenotype
MIM: 300002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARSD PubMed Links

Gene Details

Gene
ARSD
Gene Name
arylsulfatase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001669.3 4114 UTR 3 NP_001660.2
NM_009589.3 4114 Intron NP_033667.2
XM_005274514.2 4114 UTR 3 XP_005274571.1
XM_005274515.2 4114 UTR 3 XP_005274572.1

View Full Product Details