Product Details

SNP ID
rs11547662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22221803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCGTGTTGCAGCTGAAGAAGAC[C/A]CCGTGGAGCATACCGCTGGTGGGGG
Phenotype
MIM: 112264 MIM: 608511
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
BMP1 PubMed Links

Gene Details

Gene
BMP1
Gene Name
bone morphogenetic protein 1
There are no transcripts associated with this gene.

Gene
PHYHIP
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099335.1 1380 Silent Mutation GGG,GGT G181G NP_001092805.1
NM_014759.3 1380 Silent Mutation GGG,GGT G181G NP_055574.3
XM_006716416.1 1380 Silent Mutation GGG,GGT G181G XP_006716479.1
XM_017014102.1 1380 Silent Mutation GGG,GGT G48G XP_016869591.1

View Full Product Details